Diphenylphosphinic chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1415-25G C1415-5G
TCC1415-25GEA 123.09 USD
TCC1415-25G TCC1415-5G
Diphenylphosphinic chloride ≥98.0% (by GC, titration analysis)
Diphenylphosphinic chloride

CAS Number: 1499-21-4
MDL Number: MFCD00002077
Molecular Formula: C12H10ClOP
Molecular Weight: 236.63
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 26
Specific Gravity (20/20): 1.27


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Chlorodiphenylphosphine Oxide

Formula: C₁₂H₁₀ClOP
Boiling Pt: 222 °C
Density: 1.27 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00002077
CAS Number: 1499-21-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a34d2dce7aa967d9552127e6e6455842e64e025d&vid=a34d2dce7aa967d9552127e6e6455842e64e025d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR