4-Nitrophenyl chloroformate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1400-250G C1400-25G
TCC1400-250GEA 615 USD
TCC1400-250G TCC1400-25G
4-Nitrophenyl chloroformate ≥98.0% (by titrimetric analysis)
4-Nitrophenyl chloroformate

CAS Number: 7693-46-1
MDL Number: MFCD00007321
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 80
Flash Point (°C): 110
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Chloroformic Acid 4-Nitrophenyl Ester

Formula: C₇H₄ClNO₄
Boiling Pt: 162 °C
Melting Pt: 80 °C
Flash Pt: 110 °C
MDL Number: MFCD00007321
CAS Number: 7693-46-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f7d0744f4e5658edd2abb85c12bedb101ab2b0&vid=f0f7d0744f4e5658edd2abb85c12bedb101ab2b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR