(S)-(-)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-(-)-2-Chloropropanoic acid

C1373-1G C1373-25G C1373-5G
TCC1373-1GEA 17 USD
TCC1373-1G TCC1373-25G TCC1373-5G
(S)-(-)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
(S)-(-)-2-Chloropropionic acid

CAS Number: 29617-66-1
MDL Number: MFCD00064205
Molecular Formula: C3H5ClO2
Molecular Weight: 108.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.26
Specific rotation [a]20/D: -14 deg (neat)

Formula: H₃CCHClCOOH
MW: 108.52 g/mol
Density: 1.26 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00064205
CAS Number: 29617-66-1
UN: 2511
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afacb94cfcc902aae540fa0988493f38b1e6feb1&vid=afacb94cfcc902aae540fa0988493f38b1e6feb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR