3-(Chloromethyl)-3-methyloxetane ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-(Chloromethyl)-2-methyl-1,3-epoxypropane
C1229-25G
C1229-5G
TCC1229-25GEA
168
USD
TCC1229-25G
TCC1229-5G
3-(Chloromethyl)-3-methyloxetane ≥98.0% (by GC)
3-(Chloromethyl)-3-methyloxetane
CAS Number: 822-48-0
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 2-(Chloromethyl)-2-methyl-1,3-epoxypropane
Formula:
C₅H₉ClO MW: 120.58 g/mol Boiling Pt: 88 °C Density: 1.10 (20 °C) |
MDL Number:
MFCD01709552 CAS Number: 822-48-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...