2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one ≥95.0%

Supplier: TCI America

Synonyms: Van Boom's Reagent, Salicylchlorophosphite

C1210-25G C1210-5G
TCC1210-25GEA 186.2 USD
TCC1210-25G TCC1210-5G
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one ≥95.0%
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one

CAS Number: 5381-99-7
MDL Number: MFCD00013353
Molecular Formula: C7H4ClO3P
Molecular Weight: 202.53
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 39
Flash Point (°C): 110
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Salicylchlorophosphite, Van Boom's Reagent

Formula: C₇H₄ClO₃P
Melting Pt: 39 °C
Flash Pt: 110 °C
MDL Number: MFCD00013353
CAS Number: 5381-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe50ee8326e4d9d809862831c8b7a176f1b8b34a&vid=fe50ee8326e4d9d809862831c8b7a176f1b8b34a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR