3-Chlorothiophene ≥97.0% (by GC)

Supplier: TCI America

C1192-25G C1192-5G
TCC1192-25GEA 178 USD
TCC1192-25G TCC1192-5G
3-Chlorothiophene ≥97.0% (by GC)
3-Chlorothiophene

CAS Number: 17249-80-8
MDL Number: MFCD00043887
Molecular Formula: C4H3ClS
Molecular Weight: 118.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 37
Specific Gravity (20/20): 1.30

Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 139 °C
Density: 1.30 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00043887
CAS Number: 17249-80-8
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb5aad5a542c4e019ab0d1067f38061395398baf&vid=fb5aad5a542c4e019ab0d1067f38061395398baf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR