4-Cyanobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C1182-25G
C1182-5G
TCC1182-25GEA
287
USD
TCC1182-25G
TCC1182-5G
4-Cyanobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Cyanobenzoyl chloride
CAS Number: 6068-72-0
MDL Number: MFCD00001822
Molecular Formula: C8H4ClNO
Molecular Weight: 165.58
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 70
Formula:
C₈H₄ClNO MW: 165.58 g/mol Boiling Pt: 110 °C Melting Pt: 70 °C |
MDL Number:
MFCD00001822 CAS Number: 6068-72-0 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...