(±)-trans-1,2-Diaminocyclohexane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 1121-22-8
MDL Number: MFCD00063747
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 68
Freezing point (°C): 19
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: trans-1,2-Diaminocyclohexane
Formula:
C₆H₁₄N₂ Boiling Pt: 80 °C Density: 0.95 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00063747 CAS Number: 1121-22-8 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...