2-Chloropyrazine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Chloropyrazine

C1093-25G C1093-5G
TCC1093-25GEA 71.1 USD
TCC1093-25G TCC1093-5G
2-Chloropyrazine ≥98.0% (by GC)
2-Chloropyrazine

CAS Number: 14508-49-7
MDL Number: MFCD00006124
Molecular Formula: C4H3ClN2
Molecular Weight: 114.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 57
Specific Gravity (20/20): 1.30

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 93 °C
Density: 1.30 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00006124
CAS Number: 14508-49-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae26bbe8734fa62cc38c351b86f107f147b4fbf9&vid=ae26bbe8734fa62cc38c351b86f107f147b4fbf9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR