Bromocyclopropane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopropyl bromide

C1067-250G C1067-25G C1067-5G
TCC1067-250GEA 850 USD
TCC1067-250G TCC1067-25G TCC1067-5G
Bromocyclopropane ≥98.0% (by GC)
Bromocyclopropane

CAS Number: 4333-56-6
MDL Number: MFCD00001271
Molecular Formula: C3H5Br
Molecular Weight: 120.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): -6
Specific Gravity (20/20): 1.52


Other Product Names / Synonyms: Cyclopropyl Bromide

Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 69 °C
Density: 1.52 (20 °C)
Flash Pt: -6 °C
MDL Number: MFCD00001271
CAS Number: 4333-56-6
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f15d5a07146c8615adedbdbdda6be2946036f10&vid=1bfb99f57fa9b6c5da2b731da0a9922a39e9f1b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR