Cyclopropanecarbonyl chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride

C1059-25G C1059-500G
TCC1059-25GEA 55.32 USD
TCC1059-25G TCC1059-500G
Cyclopropanecarbonyl chloride ≥97.0% (by titrimetric analysis)
Cyclopropanecarbonyl chloride

CAS Number: 4023-34-1
MDL Number: MFCD00001277
Molecular Formula: C4H5ClO
Molecular Weight: 104.53
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 23
Specific Gravity (20/20): 1.15

Formula: C₄H₅ClO
MW: 104.54 g/mol
Boiling Pt: 119 °C
Density: 1.15 (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00001277
CAS Number: 4023-34-1
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7973391de20312e45399500f47d1e03fe38fe3e8&vid=7973391de20312e45399500f47d1e03fe38fe3e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR