9-(Chloromethyl)anthracene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1046-1G C1046-25G C1046-5G
TCC1046-1GEA 27 USD
TCC1046-1G TCC1046-25G TCC1046-5G
9-(Chloromethyl)anthracene ≥98.0% (by GC, titration analysis)
9-(Chloromethyl)anthracene

CAS Number: 24463-19-2
MDL Number: MFCD00001263
Molecular Formula: C15H11Cl
Molecular Weight: 226.70
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 139

Formula: C₁₅H₁₁Cl
MW: 226.71 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00001263
CAS Number: 24463-19-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405a3b530d39580e076a0e315b6cda283cfc3999&vid=eb26c843d4b935ae97b5e5ac3d64d8022a77adb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR