(1S)-(-)-Camphanic chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride

C1022-1G C1022-25G C1022-5G
TCC1022-1GEA 101 USD
TCC1022-1G TCC1022-25G TCC1022-5G
(1S)-(-)-Camphanic chloride ≥97.0% (by titrimetric analysis)
(1S)-(-)-Camphanic chloride

CAS Number: 39637-74-6
MDL Number: MFCD00135626
Molecular Formula: C10H13ClO3
Molecular Weight: 216.66
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 70
Storage Temperature: 0-10°C

Formula: C₁₀H₁₃ClO₃
MW: 216.67 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00135626
CAS Number: 39637-74-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e29c92e1cb1787c8c147c056d1c7b40605dae608&vid=e29c92e1cb1787c8c147c056d1c7b40605dae608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR