α,α,α-Trifluoro-p-toluonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 455-18-5
MDL Number: MFCD00001826
Molecular Formula: C8H4F3N
Molecular Weight: 171.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 81
Flash Point (°C): 71
Freezing point (°C): 37
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyanobenzotrifluoride, alpha,alpha,alpha-Trifluoro-p-tolunitrile
Formula:
C₈H₄F₃N Boiling Pt: 81 °C Flash Pt: 71 °C |
MDL Number:
MFCD00001826 CAS Number: 455-18-5 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...