Cyclopropanecarbonitrile ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopropyl cyanide

C0997-25ML C0997-5ML
TCC0997-25MLEA 66 USD
TCC0997-25ML TCC0997-5ML
Cyclopropanecarbonitrile ≥97.0% (by GC)
Cyclopropanecarbonitrile

CAS Number: 5500-21-0
MDL Number: MFCD00001269
Molecular Formula: C4H5N
Molecular Weight: 67.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 40
Specific Gravity (20/20): 0.90


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Cyanocyclopropane, Cyclopropanecarbonitrile

Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 135 °C
Density: 0.90 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00001269
CAS Number: 5500-21-0
UN: 3275
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f757ad091c1a28684103735b9fe684d31377fceb&vid=f757ad091c1a28684103735b9fe684d31377fceb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR