4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Chloro-2,6-dinitro-4-(trifluoromethyl)benzene, 4-Chloro-3,5-dinitrobenzotrifluoride
C0994-10G
C0994-25G
TCC0994-10GEA
71.64
USD
TCC0994-10G
TCC0994-25G
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98.0% (by GC)
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
CAS Number: 393-75-9
MDL Number: MFCD00007068
Molecular Formula: C7H2ClF3N2O4
Molecular Weight: 270.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56
Flash Point (°C): 126
Other Product Names / Synonyms: 2-Chloro-1,3-dinitro-5-(trifluoromethyl)benzene
Formula:
C₇H₂ClF₃N₂O₄ MW: 270.55 g/mol Melting Pt: 56 °C Flash Pt: 126 °C |
MDL Number:
MFCD00007068 CAS Number: 393-75-9 UN: 2811 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...