Cyclopropyl methyl ketone ≥98.0% (by GC)

Supplier: TCI America

C0985-25ML C0985-500ML C0985-100ML
TCC0985-25MLEA 32 USD
TCC0985-25ML TCC0985-500ML TCC0985-100ML
Cyclopropyl methyl ketone ≥98.0% (by GC)
Cyclopropyl methyl ketone

CAS Number: 765-43-5
MDL Number: MFCD00001297
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 13
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Acetylcyclopropane

Formula: C₅H₈O
Boiling Pt: 110 °C
Density: 0.90 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00001297
CAS Number: 765-43-5
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83d04a3f4532b09e89240c07570a7ec9929d18f&vid=d83d04a3f4532b09e89240c07570a7ec9929d18f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR