2-Chloro-3,5-dinitropyridine ≥98.0%
Supplier: TCI America
C0943-1G
C0943-5G
TCC0943-1GEA
57
USD
TCC0943-1G
TCC0943-5G
2-Chloro-3,5-dinitropyridine ≥98.0%
2-Chloro-3,5-dinitropyridine
CAS Number: 2578-45-2
MDL Number: MFCD00006233
Molecular Formula: C5H2ClN3O4
Molecular Weight: 203.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 66
Formula:
C₅H₂ClN₃O₄ Melting Pt: 66 °C |
MDL Number:
MFCD00006233 CAS Number: 2578-45-2 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...