4-Chloro-3-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride
CAS Number: 320-51-4
MDL Number: MFCD00007846
Molecular Formula: C7H5ClF3N
Molecular Weight: 195.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 36
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Amino-1-chloro-2-(trifluoromethyl)benzene, 4-Chloro-3-trifluoromethylaniline
Formula:
C₇H₅ClF₃N MW: 195.57 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00007846 CAS Number: 320-51-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...