Cyclohexyltrichlorosilane ≥98.0%
Supplier: TCI America
Synonyms:
Trichloro(cyclohexyl)silane
C0892-25G
C0892-5G
TCC0892-25GEA
150
USD
TCC0892-25G
TCC0892-5G
Cyclohexyltrichlorosilane ≥98.0%
Cyclohexyltrichlorosilane
CAS Number: 98-12-4
MDL Number: MFCD00021280
Molecular Formula: C6H11Cl3Si
Molecular Weight: 217.59
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 94
Specific Gravity (20/20): 1.23
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trichlorocyclohexylsilane
Formula:
C₆H₁₁Cl₃Si Boiling Pt: 207 °C Density: 1.23 (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00021280 CAS Number: 98-12-4 UN: 1763 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...