2-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC)
Supplier: TCI America
C0826-100G
C0826-25G
TCC0826-100GEA
404
USD
TCC0826-100G
TCC0826-25G
2-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC)
2-Chlorophenylhydrazinium chloride
CAS Number: 41052-75-9
MDL Number: MFCD00012928
Molecular Formula: C6H7ClN2
Molecular Weight: 179.04
Purity/Analysis Method:
Formula:
C₆H₈Cl₂N₂ |
MDL Number:
MFCD00012928 CAS Number: 41052-75-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...