2-Cyclopenten-1-one ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone

C0794-25ML C0794-5ML
TCC0794-25MLEA 199 USD
TCC0794-25ML TCC0794-5ML
2-Cyclopenten-1-one ≥96.0% (by GC)
2-Cyclopenten-1-one

CAS Number: 930-30-3
MDL Number: MFCD00001401
Molecular Formula: C5H6O
Molecular Weight: 82.10
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 48
Specific Gravity (20/20): 1.01
Storage Temperature: 0-10°C

Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 154 °C
Density: 1.01 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00001401
CAS Number: 930-30-3
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b10a2e102543a0fd0ca34beb2ad3cdafac2df4aa&vid=b10a2e102543a0fd0ca34beb2ad3cdafac2df4aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR