(±)-Ethyl-2-chloropropionate ≥97.0% (by GC)

Supplier: TCI America

C0790-100ML C0790-25ML
TCC0790-100MLEA 176.8 USD
TCC0790-100ML TCC0790-25ML
(±)-Ethyl-2-chloropropionate ≥97.0% (by GC)
(±)-Ethyl 2-chloropropionate

CAS Number: 535-13-7
MDL Number: MFCD00000864
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 38
Specific Gravity (20/20): 1.08


Product Categories: Synthetic Organic Chemistry, Nucleophiles (Aromatic VNS Reaction)
Other Product Names / Synonyms: 2-Chloropropionic Acid Ethyl Ester

Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 147 °C
Density: 1.08 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00000864
CAS Number: 535-13-7
UN: 2935
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6650807867da6b3279512042204db3edd8e5d47a&vid=6650807867da6b3279512042204db3edd8e5d47a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR