Cyclobutanol ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 2919-23-5
MDL Number: MFCD00001318
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 21
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: Hydroxycyclobutane
Formula:
C₄H₈O Boiling Pt: 125 °C Density: 0.92 (20 °C) Flash Pt: 21 °C |
MDL Number:
MFCD00001318 CAS Number: 2919-23-5 UN: 1120 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...