1,1-Cyclobutanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Cyclobutane-1,1-dicarboxylic acid

C0769-25G C0769-5G
TCC0769-25GEA 108.72 USD
TCC0769-25G TCC0769-5G
1,1-Cyclobutanedicarboxylic acid ≥98.0% (by GC, titration analysis)
1,1-Cyclobutanedicarboxylic acid

CAS Number: 5445-51-2
MDL Number: MFCD00001325
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 158

Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00001325
CAS Number: 5445-51-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a42bedf2922e078460da36d25d628f0c37be75f2&vid=a42bedf2922e078460da36d25d628f0c37be75f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR