α-Chloro-3-nitrotoluene ≥99.0%
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-3-nitrobenzene, 3-Nitrobenzyl chloride, α-Chloro-m-nitrotoluene, alpha-Chloro-3-nitrotoluene, m-Nitrobenzyl chloride
C0709-25G
C0709-5G
TCC0709-25GEA
147.5
USD
TCC0709-25G
TCC0709-5G
α-Chloro-3-nitrotoluene ≥99.0%
α-Chloro-3-nitrotoluene
CAS Number: 619-23-8
MDL Number: MFCD00007272
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 46
Flash Point (°C): 110
Other Product Names / Synonyms: alpha-Chloro-3-nitrotoluene
Formula:
C₇H₆ClNO₂ |
MDL Number:
MFCD00007272 CAS Number: 619-23-8 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...