Phosphonitrilic chloride trimer ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Hexachlorocyclotriphosphazene
C0584-250G
C0584-25G
TCC0584-250GEA
402.77
USD
TCC0584-250G
TCC0584-25G
Phosphonitrilic chloride trimer ≥98.0% (by GC)
Phosphonitrilic chloride trimer
CAS Number: 940-71-6
MDL Number: MFCD00006474
Molecular Formula: Cl6N3P3
Molecular Weight: 347.64
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 113
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Hexachlorocyclotriphosphazene, Cyclic Phosphonitrilic Chloride Trimer
Formula:
Cl₆N₃P₃ MW: 347.64 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD00006474 CAS Number: 940-71-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...