2-Chlorothioxanthone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one

C0292-25G C0292-5G
TCC0292-25GEA 66 USD
TCC0292-25G TCC0292-5G
2-Chlorothioxanthone ≥98.0% (by GC)
2-Chlorothioxanthone

CAS Number: 86-39-5
MDL Number: MFCD00005067
Molecular Formula: C13H7ClOS
Molecular Weight: 246.71
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152
Flash Point (°C): 91

Formula: C₁₃H₇ClOS
MW: 246.72 g/mol
Melting Pt: 152 °C
Flash Pt: 91 °C
MDL Number: MFCD00005067
CAS Number: 86-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57e7ed4c3cdb92180056125497bc8141a7b181e8&vid=57e7ed4c3cdb92180056125497bc8141a7b181e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR