α-Chloro-4-nitrotoluene ≥98.0%
Supplier: TCI America
Synonyms:
4-Nitrobenzyl chloride, 4-Nitrobenzylchloride, α-Chloro-p-nitrotoluene
C0229-250G
C0229-25G
TCC0229-250GEA
442
USD
TCC0229-250G
TCC0229-25G
α-Chloro-4-nitrotoluene ≥98.0%
α-Chloro-4-nitrotoluene
CAS Number: 100-14-1
MDL Number: MFCD00007374
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Other Product Names / Synonyms: alpha-Chloro-4-nitrotoluene
Formula:
C₇H₆ClNO₂ Melting Pt: 73 °C |
MDL Number:
MFCD00007374 CAS Number: 100-14-1 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...