Clioquinol ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 130-26-7
MDL Number: MFCD00006787
Molecular Formula: C9H5ClINO
Molecular Weight: 305.50
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: Clioquinol
Formula:
C₉H₅ClINO |
MDL Number:
MFCD00006787 CAS Number: 130-26-7 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...