1-Chloro-2,4-dinitrobenzene ≥99.0% (by GC)

Supplier: TCI America

C0162-25G C0162-500G
TCC0162-25GEA 40.31 USD
TCC0162-25G TCC0162-500G
1-Chloro-2,4-dinitrobenzene ≥99.0% (by GC)
1-Chloro-2,4-dinitrobenzene

CAS Number: 97-00-7
MDL Number: MFCD00007075
Molecular Formula: C6H3ClN2O4
Molecular Weight: 202.55
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 315
Melting point (°C): 52
Flash Point (°C): 179


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models
Other Product Names / Synonyms: 2,4-Dinitrochlorobenzene

Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 315 °C
Melting Pt: 52 °C
Flash Pt: 179 °C
MDL Number: MFCD00007075
CAS Number: 97-00-7
UN: 3441
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b253ee6d2df1b9db1cadd3d1984dc040d59b7b6a&vid=e23c6406773677e50d10db44b2ad4b74cafc69bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR