2-Bromophenethylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(2-Bromophenyl)ethylamine

B3938-25G B3938-5G
TCB3938-25GEA 355 USD
TCB3938-25G TCB3938-5G
2-Bromophenethylamine ≥98.0% (by titrimetric analysis)
2-Bromophenethylamine

CAS Number: 65185-58-2
MDL Number: MFCD01529874
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 85
Specific Gravity (20/20): 1.41


Other Product Names / Synonyms: 1-Amino-(2-bromophenyl)ethane, 2-Bromophenethylamine

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 85 °C
Density: 1.41 (20 °C)
MDL Number: MFCD01529874
CAS Number: 65185-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35b74f8a96c9e71b9e7f974cba8a63ae3f0e511e&vid=35b74f8a96c9e71b9e7f974cba8a63ae3f0e511e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR