1,2-Benzofluorene ≥95.0% (by GC)

Supplier: TCI America

B3931-1G B3931-200MG
TCB3931-1GEA 638 USD
TCB3931-1G TCB3931-200MG
1,2-Benzofluorene ≥95.0% (by GC)
1,2-Benzofluorene

CAS Number: 238-84-6
MDL Number: MFCD00039578
Molecular Formula: C17H12
Molecular Weight: 216.28
Purity/Analysis Method: >95.0% (GC)
Boiling point (°C): 407
Melting point (°C): 182
Lambda max.: 433 nm (THF)


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 11H-Benzo[a]fluorene

Formula: C₁₇H₁₂
Boiling Pt: 407 °C
Melting Pt: 182 °C
MDL Number: MFCD00039578
CAS Number: 238-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9732fa04c34d2630e1254393f61cda8eb115705a&vid=9732fa04c34d2630e1254393f61cda8eb115705a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR