Bromomethyl acetate ≥95.0% (by GC)
Supplier: TCI America
B3907-1G
B3907-5G
TCB3907-1GEA
70
USD
TCB3907-1G
TCB3907-5G
Bromomethyl acetate ≥95.0% (by GC)
Bromomethyl acetate
CAS Number: 590-97-6
MDL Number: MFCD00000170
Molecular Formula: C3H5BrO2
Molecular Weight: 152.98
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 57
Specific Gravity (20/20): 1.63
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Acetic Acid Bromomethyl Ester
Formula:
H₃CCOOCH₂Br MW: 152.98 g/mol Boiling Pt: 134 °C Density: 1.63 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00000170 CAS Number: 590-97-6 UN: 3272 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...