5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 5-Carboxybenzotriazole
B3869-25G
B3869-5G
TCB3869-25GEA
112
USD
TCB3869-25G
TCB3869-5G
5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)
5-Benzotriazolecarboxylic acid
CAS Number: 23814-12-2
MDL Number: MFCD00012318
Molecular Formula: C7H5N3O2
Molecular Weight: 163.14
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 5-Carboxybenzotriazole
Formula:
C₇H₅N₃O₂ MW: 163.14 g/mol |
MDL Number:
MFCD00012318 CAS Number: 23814-12-2 |
Specification Test Results
Purity/Analysis Method | >96.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...