Bicyclo[4.2.0]octa-1,3,5-triene ≥94.0% (by GC)
Supplier: TCI America
Synonyms:
Benzocyclobutene, Bicyclo[4.2.0]octa-1,3,5-triene
B3863-1G
B3863-200MG
TCB3863-1GEA
131.8
USD
TCB3863-1G
TCB3863-200MG
Bicyclo[4.2.0]octa-1,3,5-triene ≥94.0% (by GC)
Bicyclo[4.2.0]octa-1,3,5-triene
CAS Number: 694-87-1
MDL Number: MFCD01321219
Molecular Formula: C8H8
Molecular Weight: 104.15
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 36
Specific Gravity (20/20): 0.96
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Bicyclo[4.2.0]octa-1,3,5-triene
Formula:
C8H8 MW: 104.1500 g/mol Boiling Pt: 150 °C Density: 0.96 (20 °C) Flash Pt: 36 °C |
MDL Number:
MFCD01321219 CAS Number: 694-87-1 |
|
Specification Test Results
Purity/Analysis Method | >94.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...