(S)-(-)-Butylene oxide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(S)-(-)-1,2-Epoxybutane, (S)-(-)-Ethylethylene Oxide, (S)-(-)-Ethyloxirane
B3833-25ML
B3833-5ML
TCB3833-25MLEA
408
USD
TCB3833-25ML
TCB3833-5ML
(S)-(-)-Butylene oxide ≥98.0% (by GC)
(S)-(-)-Butylene oxide
CAS Number: 30608-62-9
MDL Number: MFCD02683445
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): -22
Specific Gravity (20/20): 0.84
Specific rotation [a]20/D: -10 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-1,2-Epoxybutane, (S)-(-)-Ethylethylene Oxide, (S)-(-)-Ethyloxirane
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 63 °C Density: 0.84 (20 °C) Flash Pt: -22 °C |
MDL Number:
MFCD02683445 CAS Number: 30608-62-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...