(R)-(+)-Butylene oxide ≥98.0%
Supplier: TCI America
Synonyms:
(R)-(+)-Ethylethylene Oxide, (R)-(+)-Ethyloxirane, (R)-(+)-1,2-Epoxybutane
B3832-25ML
B3832-5ML
TCB3832-25MLEA
279.3
USD
TCB3832-25ML
TCB3832-5ML
(R)-(+)-Butylene oxide ≥98.0%
(R)-(+)-Butylene oxide
CAS Number: 3760-95-0
MDL Number: MFCD02683444
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): -22
Specific Gravity (20/20): 0.84
Specific rotation [a]20/D: 10 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-1,2-Epoxybutane, (R)-(+)-Ethylethylene Oxide, (R)-(+)-Ethyloxirane
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 63 °C Density: 0.84 (20 °C) Flash Pt: –22 °C |
MDL Number:
MFCD02683444 CAS Number: 3760-95-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...