5-Bromo-o-anisidine ≥97.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

B3828-1G B3828-5G
TCB3828-1GEA 32 USD
TCB3828-1G TCB3828-5G
5-Bromo-o-anisidine ≥97.0%
5-Bromo-o-anisidine

CAS Number: 6358-77-6
MDL Number: MFCD04037882
Molecular Formula: C7H8BrNO
Molecular Weight: 202.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: 2-Amino-4-bromoanisole, 5-Bromo-o-anisidine

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 95 °C
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f4c540c456208422e9c64589a8511f702b92e07&vid=4f4c540c456208422e9c64589a8511f702b92e07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR