Benzyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis) for biochemical research
Supplier: TCI America
Synonyms:
Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate
B3768-25G
B3768-5G
TCB3768-25GEA
31.34
USD
TCB3768-25G
TCB3768-5G
Benzyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis) for biochemical research
Benzyl-4-hydroxybenzoate
[for Biochemical Research]
CAS Number: 94-18-8
MDL Number: MFCD00016471
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113
Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Benzyl Ester, Benzylparaben
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD00016471 CAS Number: 94-18-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...