Butyl isovalerate ≥98.0%
Supplier: TCI America
Synonyms:
Isovaleric acid butyl ester, Butyl 3-methylbutanoate
B3718-25ML
B3718-500ML
TCB3718-25MLEA
26
USD
TCB3718-25ML
TCB3718-500ML
Butyl isovalerate ≥98.0%
Butyl isovalerate
CAS Number: 109-19-3
MDL Number: MFCD00048775
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Flash Point (°C): 53
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Isovaleric Acid Butyl Ester
Formula:
C₉H₁₈O₂ Boiling Pt: 176 °C Density: 0.86 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00048775 CAS Number: 109-19-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...