5-Bromonicotinonitrile ≥98.0%
Supplier: TCI America
Synonyms:
5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine
B3713-1G
B3713-5G
TCB3713-1GEA
35.6
USD
TCB3713-1G
TCB3713-5G
5-Bromonicotinonitrile ≥98.0%
5-Bromonicotinonitrile
CAS Number: 35590-37-5
MDL Number: MFCD00174363
Molecular Formula: C6H3BrN2
Molecular Weight: 183.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105
Other Product Names / Synonyms: 3-Bromo-5-cyanopyridine, 5-Bromonicotinonitrile
Formula:
C₆H₃BrN₂ MW: 183.01 Melting Pt: 105 °C |
MDL Number:
MFCD00174363 CAS Number: 35590-37-5 UN: 3439 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...