(S)-(-)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-(4-Bromophenyl)ethanamine, (S)-(-)-1-(4-Bromophenyl)ethylamine

B3674-25G B3674-5G
TCB3674-25GEA 658.85 USD
TCB3674-25G TCB3674-5G
(S)-(-)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-4-Bromo-α-methylbenzylamine

CAS Number: 27298-97-1
MDL Number: MFCD00066026
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.39
Specific rotation [a]20/D: -18 deg (C=3, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-4-Bromo-alpha-methylbenzylamine

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Density: 1.39 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00066026
CAS Number: 27298-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f661b086690ddbbcfd57f50dd60555a7b82de49&vid=2f661b086690ddbbcfd57f50dd60555a7b82de49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR