4-Bromo-4'-iodobiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-4'-iodo-1,1'-biphenyl, 1-Bromo-4-(4-iodophenyl)benzene

B3648-25G B3648-5G
TCB3648-25GEA 75.3 USD
TCB3648-25G TCB3648-5G
4-Bromo-4'-iodobiphenyl ≥98.0% (by GC)
4-Bromo-4'-iodobiphenyl

CAS Number: 105946-82-5
Molecular Formula: C12H8BrI
Molecular Weight: 359.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 177

Formula: C₁₂H₈BrI
MW: 359.00 g/mol
Melting Pt: 177 °C
MDL Number: MFCD17676204
CAS Number: 105946-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1eece6992e85c4c31a30faafa643264116d9e89&vid=f1eece6992e85c4c31a30faafa643264116d9e89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR