3,5-Bis(trifluoromethyl)benzyl chloride ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Chloromethyl)-3,5-bis(trifluoromethyl)benzene

B3641-25G B3641-5G
TCB3641-25GEA 179 USD
TCB3641-25G TCB3641-5G
3,5-Bis(trifluoromethyl)benzyl chloride ≥97.0% (by GC)
3,5-Bis(trifluoromethyl)benzyl chloride

CAS Number: 75462-59-8
MDL Number: MFCD00009906
Molecular Formula: C9H5ClF6
Molecular Weight: 262.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 71


Other Product Names / Synonyms: 1-(Chloromethyl)-3,5-bis(trifluoromethyl)benzene

Formula: C₉H₅ClF₆
MW: 262.58 g/mol
Boiling Pt: 84 °C
Flash Pt: 71 °C
MDL Number: MFCD00009906
CAS Number: 75462-59-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7877c7125dc7aaa28c2b4339168e494c2d6ef2bc&vid=7877c7125dc7aaa28c2b4339168e494c2d6ef2bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR