trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
DCTB
B3635-100MG
B3635-1G
TCB3635-100MGEA
33.05
USD
TCB3635-100MG
TCB3635-1G
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile ≥98.0% (by HPLC, total nitrogen)
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile
CAS Number: 300364-84-5
MDL Number: MFCD03453260
Molecular Formula: C17H18N2
Molecular Weight: 250.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 125
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: DCTB
Formula:
C₁₇H₁₈N₂ MW: 250.35 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD03453260 CAS Number: 300364-84-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...