5-Bromopyridine-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America

B3610-25G B3610-5G
TCB3610-25GEA 133 USD
TCB3610-25G TCB3610-5G
5-Bromopyridine-2-carbonitrile ≥98.0% (by GC)
5-Bromopyridine-2-carbonitrile

CAS Number: 97483-77-7
MDL Number: MFCD00234144
Molecular Formula: C6H3BrN2
Molecular Weight: 183.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 131


Other Product Names / Synonyms: 5-Bromo-2-pyridinecarbonitrile, 5-Bromopicolinonitrile

Formula: C₆H₃BrN₂
Boiling Pt: 110 °C
Melting Pt: 131 °C
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72307892ac2d7a2ea02ca86aeff1c562262819c5&vid=72307892ac2d7a2ea02ca86aeff1c562262819c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR