(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)

Supplier: TCI America

B3592-1G B3592-200MG
TCB3592-1GEA 419.5 USD
TCB3592-1G TCB3592-200MG
(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate

CAS Number: 862095-77-0 Molecular Formula: C19H17BF7N3 Molecular Weight: 431.16 Purity/Analysis Method: 97.0% (N) Form: Crystal Melting point (°C): 203

Formula: C₁₉H₁₇BF₇N₃
MW: 431.16 g/mol
Melting Pt: 203 °C
CAS Number: 862095-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21a093aa81e4d842470aa7a66be612e7d7f84b34&vid=21a093aa81e4d842470aa7a66be612e7d7f84b34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR