α-Bromo-m-xylene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Methylbenzyl bromide

B3585-25G B3585-5G
TCB3585-25GEA 1018 USD
TCB3585-25G TCB3585-5G
α-Bromo-m-xylene ≥98.0% (by GC)
α-Bromo-m-xylene

CAS Number: 620-13-3
MDL Number: MFCD00000177
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 82
Specific Gravity (20/20): 1.37


Other Product Names / Synonyms: 3-Methylbenzyl Bromide

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 98 °C
Density: 1.37 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00000177
CAS Number: 620-13-3
UN: 1701
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ddad88478673be8669d24a53406580e4950867b&vid=59c3a5c9aa76ef63522203065edfa076a3e59fb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR