tert-Butyl propionate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Propionic Acid tert-Butyl Ester

B3327-100G B3327-25G
TCB3327-100GEA 277 USD
TCB3327-100G TCB3327-25G
tert-Butyl propionate ≥98.0% (by GC)
tert-Butyl propionate

CAS Number: 20487-40-5
MDL Number: MFCD00009304
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 21
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Propionic Acid tert-Butyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 119 °C
Density: 0.86 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00009304
CAS Number: 20487-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19aca772570ab4a27531807752b547342923b4f1&vid=19aca772570ab4a27531807752b547342923b4f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR